missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylcyclohexanone, 99%

Product Code. p-7081234
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11409023

Brand: Thermo Scientific Alfa Aesar A11126.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40649-36-3 | C9H16O | 140.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40649-36-3
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00058690
InChI Key NQEDLIZOPMNZMC-UHFFFAOYSA-N
Synonym 4-propylcyclohexanone, 4-n-propylcyclohexanone, cyclohexanone, 4-propyl, 4-propyl-1-cyclohexanone, 4-propylcyclohexanon, 4-propyl-cyclohexanone, pubchem14023, acmc-1arvj, 4beta-propylcyclohexanone, 4-n-propyl-cyclohexanone
PubChem CID 142482
IUPAC Name 4-propylcyclohexan-1-one
SMILES CCCC1CCC(=O)CC1

Specifications

Density 0.91
Boiling Point 115°C (36 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.4535
Quantity 25 g
Beilstein 2076023
Formula Weight 140.23
Percent Purity 99%
Chemical Name or Material 4-n-Propylcyclohexanone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.