missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-3-pyrazolecarboxylic acid, 95%

Product Code. 10795632 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10795632 10 g Glass bottle 10g
10432980 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10795632 Supplier Thermo Scientific Acros Supplier No. 427540100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5334-40-7 | C4H3N3O4 | 157.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5334-40-7
Molecular Formula C4H3N3O4
Molecular Weight (g/mol) 157.08
MDL Number MFCD00090899
InChI Key ZMAXXOYJWZZQBK-UHFFFAOYSA-N
Synonym 4-nitro-1h-pyrazole-3-carboxylic acid, 4-nitropyrazole-3-carboxylic acid, 4-nitro-3-pyrazolecarboxylic acid, 4-nitro-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 4-nitro, 4-nitro-2h-pyrazole-3-carboxylic acid, 4-nitro pyrazole-3-carboxylic acid, 4-nitro-pyrazole-3-carboxylic acid, 4-hydroxynitroso-1h-pyrazole-3-carboxylic acid, nitropyrazolecarboxylic
PubChem CID 219739
SMILES C1=NNC(=C1[N+](=O)[O-])C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 223.0°C to 225.0°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 157.08
Percent Purity 95%
Chemical Name or Material 4-Nitro-3-pyrazolecarboxylic acid
compliance-icons
Product Identifier
  • 4-Nitro-3-pyrazolecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.