missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-o-phenylenediamine, 97%

Product Code. 11321677
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11321677 100 g 100g
11331677 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11321677 Supplier Thermo Scientific Alfa Aesar Supplier No. L04693.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 99-56-9 | C6H7N3O2 | 153.14 g/mol

4-Nitro-o-phenylenediamine is used to study the in vitro activation of 4-nitro-o-phenylenediamine into a potent mutagen in Salmonella typhimurium strain TA98

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitro-o-phenylenediamine is used to study the in vitro activation of 4-nitro-o-phenylenediamine into a potent mutagen in Salmonella typhimurium strain TA98

Solubility
Slightly soluble in water (1.2 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-56-9
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007724
InChI Key RAUWPNXIALNKQM-UHFFFAOYSA-N
Synonym 4-nitro-o-phenylenediamine, 1,2-diamino-4-nitrobenzene, 4-nitro-1,2-phenylenediamine, 2-amino-4-nitroaniline, 3,4-diaminonitrobenzene, p-nitro-o-phenylenediamine, 1,2-benzenediamine, 4-nitro, 4-nitro-1,2-diaminobenzene, 4-nitrophenylenediamine, 4-nitro-1,2-benzenediamine
PubChem CID 5111791
ChEBI CHEBI:67116
IUPAC Name 4-nitrobenzene-1,2-diamine
SMILES NC1=CC=C(C=C1N)[N+]([O-])=O

Specifications

Melting Point 199°C to 203°C
Quantity 100 g
Beilstein 608106
Merck Index 14,6623
Solubility Information Slightly soluble in water (1.2g/L at 20°C).
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 4-Nitro-o-phenylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.