missing translation for 'onlineSavingsMsg'
Learn More

4'-Nitroacetophenone, 98%

Product Code. 11431568
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431568 250 g 250g
11421568 50 g 50g
11411568 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431568 Supplier Thermo Scientific Alfa Aesar Supplier No. A13877.30
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-19-6 | C8H7NO3 | 165.15 g/mol

4'-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, strong alkalis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-19-6
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00007355
InChI Key YQYGPGKTNQNXMH-UHFFFAOYSA-N
Synonym p-nitroacetophenone, 4'-nitroacetophenone, 4-nitroacetophenone, 1-4-nitrophenyl ethanone, p-acetylnitrobenzene, paranitroacetophenone, p-nitrophenyl methyl ketone, 1-4-nitrophenyl ethan-1-one, ethanone, 1-4-nitrophenyl, pnap
PubChem CID 7487
ChEBI CHEBI:28735
IUPAC Name 1-(4-nitrophenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 77°C to 81°C
Boiling Point 201°C to 202°C
Flash Point 149°C (300°F)
Assay Percent Range 98%
Odor Characteristic
Quantity 250 g
Beilstein 607777
Solubility Information Insoluble in water.
Formula Weight 165.15
Percent Purity 98%
Chemical Name or Material 4'-Nitroacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.