missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroaniline, 99%

Product Code. 11935211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11935211 2.5 kg Plastic Bottle 2.5kg
10657794 100 g Glass bottle 100g
10523871 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11935211 Supplier Thermo Scientific Acros Supplier No. 128370025
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-01-6 | C6H6N2O2 | 138.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-01-6
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00007858
InChI Key TYMLOMAKGOJONV-UHFFFAOYSA-N
Synonym p-nitroaniline, p-nitraniline, 4-nitrobenzenamine, p-aminonitrobenzene, 4-nitraniline, benzenamine, 4-nitro, p-nitrophenylamine, 1-amino-4-nitrobenzene, aniline, p-nitro, developer p
PubChem CID 7475
ChEBI CHEBI:17064
IUPAC Name 4-nitroaniline
SMILES NC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 146.0°C to 151.0°C
Color Yellow to Brown
Boiling Point 332.0°C
Flash Point 199°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic Bottle
Linear Formula O2NC6H4NH2
Quantity 2.5 kg
Beilstein 12,711
Merck Index 15,667
Solubility Information Solubility in water: 0.8g/L (20°C). Other solubilities: soluble in alcohol,ether,benzene and methanol,1 g soluble in 25 mL alcohol,1 g soluble in 30 mL ether
Formula Weight 138.13
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 4-Nitroaniline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Nitroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: p-Nitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.