missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroaniline, TRC

Product Code. 30472356
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30472356 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30472356 Supplier Toronto Research Chemicals Supplier No. TRCN4917705G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-01-6
Molecular Formula C6 H6 N2 O2
Molecular Weight (g/mol) 138.12
Synonym Benzenamine, 4-nitro-, Aniline, p-nitro- (8CI), 4-Nitrobenzenamine, 1-Amino-4-nitrobenzene, 4-Amino-1-nitrobenzene, 4-Aminonitrobenzene, 4-Nitro-1-aminobenzene, 4-Nitroaniline, 4-Nitrophenylamine, Azoamine Red Zh, C.I. 37035, C.I. Azoic Diazo Component 37, C.I. Developer 17, Developer P, Devol Red GG, Fast Red 2G Base, Fast Red Base 2J, Fast Red Base GG, Fast Red GG Base, Fast Red MP Base, Fast Red P Base, NSC 9797, Naphtoelan Red GG Base, Nitrazol CF extra, PNA, Red 2G Base, Shinnippon Fast Red GG Base, p-Aminonitrobenze, p-Aminonitrobenzene, p-Nitraniline, p-Nitroaniline, p-Nitrophenylamine
IUPAC Name 4-nitroaniline
SMILES Nc1ccc(cc1)[N+](=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H6N2O2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H,7H2
Formula Weight 138.0429
Chemical Name or Material 4-Nitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.