missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzenediazonium tetrafluoroborate, 97%

Product Code. 15431538
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
15431538 1 g 1g
15441538 5 g 5g
15451538 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 15431538

Brand: Thermo Scientific Alfa Aesar H55315.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 456-27-9 | C6H4BF4N3O2 | 236.92 g/mol

4-Nitrobenzenediazonium tetrafluoroborate is widely used as a standard reagent in organic synthesis. It is used as a dye for enterochromaffin cells. It is used for the spectrophotometric determination of common aromatic diisocyanates [toluene-2,4-diyl diisocyanate and 4,4'-methylenebis(phenyl isocyanate)] and gallic acid. It is also used for tissue staining and determination of trace phenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzenediazonium tetrafluoroborate is widely used as a standard reagent in organic synthesis. It is used as a dye for enterochromaffin cells. It is used for the spectrophotometric determination of common aromatic diisocyanates [toluene-2,4-diyl diisocyanate and 4,4′-methylenebis(phenyl isocyanate)] and gallic acid. It is also used for tissue staining and determination of trace phenols.

Solubility
Soluble in acetonitrile and dimethyl sulfoxide.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed. Incompatible with strong bases, strong oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-27-9
Molecular Formula C6H4BF4N3O2
Molecular Weight (g/mol) 236.92
MDL Number MFCD00012005
InChI Key AFULQCYCQAHYIP-UHFFFAOYSA-N
Synonym 4-nitrobenzenediazonium tetrafluoroborate, ccris 3641, unii-2uxo615r4o, p-nitrobenzenediazonium tetrafluoroborate, benzenediazonium, 4-nitro-, tetrafluoroborate 1-, p-nitrophenyldiazonium fluoborate, p-nitrobenzenediazonium fluoborate, 4-nitrobenzenediazonium fluoborate, p-nitrobenzenediazonium fluoroborate, 4-nitrobenzenediazonium fluoroborate
PubChem CID 68006
IUPAC Name 4-nitrobenzenediazonium;tetrafluoroborate
SMILES F[B-](F)(F)F.[O-][N+](=O)C1=CC=C(C=C1)[N+]#N

Specifications

Melting Point 144°C to 148°C
Quantity 1 g
UN Number UN2923
Beilstein 3599869
Solubility Information Soluble in acetonitrile and dimethyl sulfoxide.
Formula Weight 236.92
Percent Purity 97%
Chemical Name or Material 4-Nitrobenzenediazonium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.