missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzenesulfonyl chloride, 95%

Product Code. 10238083 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10238083 5 g Glass bottle 5g
10791442 25 g Glass bottle 25g
10290031 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10238083 Supplier Thermo Scientific Acros Supplier No. 415850050
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-74-8 | C6H4ClNO4S | 221.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-74-8
Molecular Formula C6H4ClNO4S
Molecular Weight (g/mol) 221.61
MDL Number MFCD00007445
InChI Key JXRGUPLJCCDGKG-UHFFFAOYSA-N
Synonym 4-nitrobenzenesulfonylchloride, 4-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-nitro, 4-nitrobenzenesulphonyl chloride, 4-nitrophenylsulfonyl chloride, p-nitrophenylsulfonyl chloride, 4-nitrobenzenesulfonic acid chloride, p-nitrobenzenesulfonyl chloride, 4-nitrobenzenesulphonylchloride, 4-nitro-benzenesulfonyl chloride
PubChem CID 7404
IUPAC Name 4-nitrobenzenesulfonyl chloride
SMILES [O-][N+](=O)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 75°C to 80°C
Color Yellow to Brown
Boiling Point 143°C to 144°C (2.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula SO2ClC6H4NO2
Quantity 5 g
Beilstein 11, 72
Solubility Information Solubility in water: insoluble
Formula Weight 221.62
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Nitrobenzenesulfonyl chloride, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.