missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzenesulfonyl chloride, 95%

Product Code. 10290031 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10290031

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-74-8 | C6H4ClNO4S | 221.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-74-8
Molecular Formula C6H4ClNO4S
Molecular Weight (g/mol) 221.61
MDL Number MFCD00007445
InChI Key JXRGUPLJCCDGKG-UHFFFAOYSA-N
Synonym 4-nitrobenzenesulfonylchloride, 4-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-nitro, 4-nitrobenzenesulphonyl chloride, 4-nitrophenylsulfonyl chloride, p-nitrophenylsulfonyl chloride, 4-nitrobenzenesulfonic acid chloride, p-nitrobenzenesulfonyl chloride, 4-nitrobenzenesulphonylchloride, 4-nitro-benzenesulfonyl chloride
PubChem CID 7404
IUPAC Name 4-nitrobenzenesulfonyl chloride
SMILES [O-][N+](=O)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 75°C to 80°C
Color Yellow to Brown
Boiling Point 143°C to 144°C (2.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula SO2ClC6H4NO2
Quantity 100 g
Beilstein 11, 72
Solubility Information Solubility in water: insoluble
Formula Weight 221.62
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Nitrobenzenesulfonyl chloride, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.