missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzhydrazide, 98+%

Product Code. 11499146
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499146 10 g 10g
11409156 50 g 50g
11419156 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499146 Supplier Thermo Scientific Alfa Aesar Supplier No. A18016.09
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 636-97-5 | C7H7N3O3 | 181.151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-97-5
Molecular Formula C7H7N3O3
Molecular Weight (g/mol) 181.151
MDL Number MFCD00007604
InChI Key FKZXYJYTUSGIQE-UHFFFAOYSA-N
Synonym 4-nitrobenzhydrazide, p-nitrobenzhydrazide, 4-nitrobenzoylhydrazine, p-nitrobenzohydrazide, p-nitrobenzoylhydrazide, p-nitrobenzoic hydrazide, p-nitrobenzoic acid hydrazide, p-nitrobenzoyl hydrazine, 4-nitrobenzoic acid hydrazide, 4-nitrobenzoyl hydrazide
PubChem CID 3693744
ChEBI CHEBI:66928
IUPAC Name 4-nitrobenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)[N+](=O)[O-]

Specifications

Melting Point ∼216°C (decomposition)
Quantity 10 g
Beilstein 519882
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material 4-Nitrobenzhydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.