missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzonitrile, 97%

Product Code. 10677092 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10677092 25 g Glass bottle 25g
13448338 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10677092 Supplier Thermo Scientific Acros Supplier No. 128480250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 619-72-7 | C7H4N2O2 | 148.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-72-7
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.12
MDL Number MFCD00007279
InChI Key NKJIFDNZPGLLSH-UHFFFAOYSA-N
Synonym benzonitrile, 4-nitro, 4-cyanonitrobenzene, p-nitrobenzonitrile, p-cyanonitrobenzene, benzonitrile, p-nitro, 4-nitrobenzenecarbonitrile, 1-nitro-4-cyanobenzene, 4-nitro-benzonitrile, ccris 2328, 4-nitrobenzonitril
PubChem CID 12090
IUPAC Name 4-nitrobenzonitrile
SMILES [O-][N+](=O)C1=CC=C(C=C1)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 144.0°C to 149.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4CN
Quantity 25 g
Beilstein 09,397
Solubility Information Solubility in water: sparingly soluble
Formula Weight 148.12
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Nitrobenzonitrile, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Nitrobenzonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.