missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl Bromide, TRC

Product Code. 30476823
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30476823 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30476823 Supplier Toronto Research Chemicals Supplier No. TRCN49364025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Nitrobenzyl Bromide
CAS 100-11-8
Molecular Formula C7H6BrNO2
Quantity 25 g
Synonym 1-(Bromomethyl)-4-nitrobenzene, alpha-Bromo-p-nitro-toluene, 1-(Bromomethyl)-4-nitrobenzene, 4-(Bromomethyl)nitrobenzene, 4-Nitrobenzyl Bromide, 4-Nitrophenylmethyl Bromide, NSC 4609, p-(Bromomethyl)nitrobenzene, p-Nitro-alpha-bromotoluene, p-Nitrobenzyl Bromide, alpha-Bromo-4-nitrotoluene, alpha-Bromo-p-nitrotoluene
InChI Formula InChI=1S/C7H6BrNO2/c8-5-6-1-3-7(4-2-6)9(10)11/h1-4H,5H2
SMILES [O-][N+](=O)c1ccc(CBr)cc1
IUPAC Name 1-(bromomethyl)-4-nitrobenzene
Molecular Weight (g/mol) 216.03
Formula Weight 214.9582
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.