missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl Chloroformate, TRC

Product Code. 30472105
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30472105 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30472105 Supplier Toronto Research Chemicals Supplier No. TRCN49364525G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Nitrobenzyl Chloroformate
CAS 4457-32-3
Molecular Formula C8H6ClNO4
Quantity 25 g
Synonym Chloroformic Acid p-Nitrobenzyl Ester, 4-Nitrobenzyl Chloroformate, 4-Nitrobenzyloxycarbonyl Chloride, p-Nitrobenzyl Chloroformate, p-Nitrobenzyloxycarbonyl Chloride, p-Nitrocarbobenzoxy Chloride
InChI Formula InChI=1S/C8H6ClNO4/c9-8(11)14-5-6-1-3-7(4-2-6)10(12)13/h1-4H,5H2
SMILES [O-][N+](=O)c1ccc(COC(=O)Cl)cc1
IUPAC Name (4-nitrophenyl)methyl carbonochloridate
Molecular Weight (g/mol) 215.69
Formula Weight 214.9985
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.