missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrodiphenylamine, 98+%

Product Code. 11390184
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11390184 5 g 5g
11380184 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11390184 Supplier Thermo Scientific Alfa Aesar Supplier No. L00728.06
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 836-30-6 | C12H10N2O2 | 214.22 g/mol

4-Nitrodiphenylamine, is used as a Hammett indicator. It is also used as an Intermediate in the production of rubber antioxidants. It is manufactured solely for use as a chemical intermediate for industrial uses and thus is essentially fully consumed in chemical reductive alkylation reaction downstream of manufacture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrodiphenylamine, is used as a Hammett indicator. It is also used as an Intermediate in the production of rubber antioxidants. It is manufactured solely for use as a chemical intermediate for industrial uses and thus is essentially fully consumed in chemical reductive alkylation reaction downstream of manufacture.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 836-30-6
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.22
MDL Number MFCD00007301
InChI Key XXYMSQQCBUKFHE-UHFFFAOYSA-N
Synonym 4-nitrodiphenylamine, p-nitrodiphenylamine, benzenamine, 4-nitro-n-phenyl, diphenylamine, 4-nitro, 4-nitrodifenylamin, 4-ndpa, p-nitrophenylphenylamine, 4-nitro-n-phenyl-aniline, 4-nitrophenyl phenylamine, 4-nitrodifenylamin czech
PubChem CID 13271
IUPAC Name 4-nitro-N-phenylaniline
SMILES [O-][N+](=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point 132°C to 135°C
Boiling Point 211°C (30 mmHg)
Flash Point 190°C (374°F)
Quantity 5 g
UN Number UN3077
Beilstein 2051910
Solubility Information Insoluble in water.
Formula Weight 214.22
Percent Purity ≥98%
Chemical Name or Material 4-Nitrodiphenylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.