missing translation for 'onlineSavingsMsg'
Learn More

Melford 4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside

Product Code. 30018594
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018594 1 g 1g
30018593 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018594 Supplier Melford Supplier No. N516001.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chromogenic substrate for detecting N-Acetyl-β-D-glucosaminidase.

Chromogenic substrate for detecting N-Acetyl-β-D-glucosaminidase.

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.