missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl b-D-Galactofuranoside, TRC

Product Code. 30470297
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30470297 25 mg -
30469145 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30470297 Supplier Toronto Research Chemicals Supplier No. TRCN50357025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 100645-45-2
Molecular Formula C12 H15 N O8
Molecular Weight (g/mol) 301.25
Synonym β-D-Galactofuranoside, 4-nitrophenyl, 4-Nitrophenyl β-D-galactofuranoside, p-Nitrophenyl β-D-galactofuranoside
IUPAC Name (2S,3R,4R,5S)-2-[(1R)-1,2-dihydroxyethyl]-5-(4-nitrophenoxy)oxolane-3,4-diol
SMILES OC[C@@H](O)[C@@H]1O[C@@H](Oc2ccc(cc2)[N+](=O)[O-])[C@H](O)[C@H]1O

Specifica

Quantity 25 mg
InChI Formula InChI=1S/C12H15NO8/c14-5-8(15)11-9(16)10(17)12(21-11)20-7-3-1-6(2-4-7)13(18)19/h1-4,8-12,14-17H,5H2/t8-,9-,10-,11+,12-/m1/s1
Formula Weight 301.0798
Chemical Name or Material 4-Nitrophenyl beta-D-Galactofuranoside
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.