missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl-beta-D-galactopyranoside hydrate, 98+%

Product Code. 11313436
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11313436 1 g 1g
11323436 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11313436

Brand: Thermo Scientific Alfa Aesar L12911.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 200422-18-0 | C12H15NO8 | 301.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 200422-18-0
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.25
MDL Number MFCD00063256
InChI Key IFBHRQDFSNCLOZ-MFZRPPIDNA-N
Synonym 4-nitrophenyl-beta-d-galactopyranoside hydrate, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-4-nitrophenoxy tetrahydro-2h-pyran-3,4,5-triol hydrate, p-nitrophenyl-?-d-galactoside hydrate, 4-nitrophenyl beta-d-galactopyranoside hydrate
PubChem CID 16218614
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol;hydrate
SMILES OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Melting Point 180°C to 184°C
Quantity 1 g
Beilstein 92213
Optical Rotation −84° (c=0.3 in Water)
Formula Weight 319.27 (301.25 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 4-Nitrophenyl-beta-D-galactopyranoside hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.