missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl-beta-D-glucopyranoside, 98+%

Product Code. 11378265
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378265 1 g 1g
11388265 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378265 Supplier Thermo Scientific Alfa Aesar Supplier No. L10154.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2492-87-7 | C12H15NO8 | 301.25 g/mol

4-Nitrophenyl-β-D-glucopyranoside acts as a chromogenic beta-D-glucosidase substrate used for the measurement of beta-glucosidase activity. Further, it is used in research, biochemical enzyme assays and in vitro diagnostic analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl-β-D-glucopyranoside acts as a chromogenic beta-D-glucosidase substrate used for the measurement of beta-glucosidase activity. Further, it is used in research, biochemical enzyme assays and in vitro diagnostic analysis.

Solubility
Soluble in water, methanol, warm ethanol, dimethyl sulfoxide and dimethylformamide. Insoluble in ether.

Notes
Store in a cool place. Incompatible with strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2492-87-7
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.25
MDL Number MFCD00006593
InChI Key IFBHRQDFSNCLOZ-RMPHRYRLSA-N
Synonym 4-nitrophenyl-beta-d-glucopyranoside, pnpg, 4-nitrophenyl beta-d-glucopyranoside, 1-o-p-nitrophenyl-d-glucose, 4-nitrophenyl beta-d-glucoside, p-nitrophenyl beta-d-glucopyranoside, p-nitrophenyl beta-d-glucoside, glucoside, p-nitrophenyl, p-nitrophenyl-beta-glucoside, p-nitrophenyl beta-glucoside
PubChem CID 92930
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 166°C to 167°C
Quantity 1 g
Beilstein 92211
Solubility Information Soluble in water,methanol,warm ethanol,dimethyl sulfoxide and dimethylformamide. Insoluble in ether.
Optical Rotation −103° (c=1 in Water)
Formula Weight 301.25
Percent Purity ≥98%
Chemical Name or Material 4-Nitrophenyl-beta-D-glucopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.