missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl-beta-D-Glucopyranoside

Product Code. 15855178 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15855178 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15855178 Supplier Thermo Scientific Supplier No. J19594.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2492-87-7 | C12H15NO8 | 301.251 g/mol

 

TRUSTED_SUSTAINABILITY

Specifications

CAS 2492-87-7
Melting Point 166°C to 167°C
Packaging Plastic bottle
Molecular Formula C12H15NO8
MDL Number MFCD00006593
Quantity 5 g
Beilstein 92211
Synonym 4-nitrophenyl-beta-d-glucopyranoside, pnpg, 4-nitrophenyl beta-d-glucopyranoside, 1-o-p-nitrophenyl-d-glucose, 4-nitrophenyl beta-d-glucoside, p-nitrophenyl beta-d-glucopyranoside, p-nitrophenyl beta-d-glucoside, glucoside, p-nitrophenyl, p-nitrophenyl-beta-glucoside, p-nitrophenyl beta-glucoside
InChI Key IFBHRQDFSNCLOZ-RMPHRYRLSA-N
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O
IUPAC Name (2 R,3 S,4 S,5 R,6 S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular Weight (g/mol) 301.251
PubChem CID 92930
Formula Weight 301.25
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl-beta-D-Glucopyranoside
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.