missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl formate, 97%, Thermo Scientific™

Product Code. 11974891 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11974891 25 g 25g
11342640 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11974891 Supplier Thermo Scientific Acros Supplier No. 438780250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1865-01-6
Molecular Weight (g/mol) 167.12
InChI Key IEXRKQFZXJSHOB-UHFFFAOYSA-N
PubChem CID 74628
IUPAC Name (4-nitrophenyl) formate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 66.0°C to 73.0°C
Color White
Elemental Analysis 48.30 to 52.32% C, 8.05 to 8.72% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C7H5NO4
Quantity 25 g
Synonym p-nitrophenyl formate, formic acid, 4-nitrophenyl ester, 4-nitrophenyl formate, formic acid, p-nitrophenyl ester, formic acid 4-nitrophenyl ester, acmc-1bty1, formic acid,4-nitrophenyl ester, formic acid 4-nitro-phenyl ester, 4-nitrophenyl formate hplc
Formula Weight 167.12
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Nitrophenyl formate
Show More Show Less
Product Identifier
  • 4-Nitrophenyl formate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.