missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl formate, 98%

Product Code. 11389993
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11389993 25 g 25g
11379993 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389993 Supplier Thermo Scientific Alfa Aesar Supplier No. L00316.14
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1865-01-6 | C7H5NO4 | 167.12 g/mol

Convenient and selective reagent for the selective formylation of terminal amino groups in such basic amino acids as ornithine or lysine and formylation of α-amino groups in peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Convenient and selective reagent for the selective formylation of terminal amino groups in such basic amino acids as ornithine or lysine and formylation of α-amino groups in peptides.

Solubility
Hydrolyzes in water. Soluble in dichloromethane.

Notes
Moisture and light sensitive. Store away from water/moisture, oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark. Store under inert gas. Protect from exposure to light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1865-01-6
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00014713
InChI Key IEXRKQFZXJSHOB-UHFFFAOYSA-N
PubChem CID 74628
IUPAC Name (4-nitrophenyl) formate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC=O

Specifications

Melting Point 70°C to 74°C
Quantity 25 g
Beilstein 1950897
Sensitivity Light and moisture sensitive
Synonym p-nitrophenyl formate, formic acid, 4-nitrophenyl ester, 4-nitrophenyl formate, formic acid, p-nitrophenyl ester, formic acid 4-nitrophenyl ester, acmc-1bty1, formic acid,4-nitrophenyl ester, formic acid 4-nitro-phenyl ester, 4-nitrophenyl formate hplc
Solubility Information Hydrolyzes in water. Soluble in dichloromethane.
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material 4-Nitrophenyl formate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.