missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl phenyl sulfide, 98%

Product Code. 11443507
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443507 25 g 25g
11433507 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443507 Supplier Thermo Scientific Alfa Aesar Supplier No. A17278.14
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 952-97-6 | C12H9NO2S | 231.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 952-97-6
Molecular Formula C12H9NO2S
Molecular Weight (g/mol) 231.27
MDL Number MFCD00024700
InChI Key RJCBYBQJVXVVKB-UHFFFAOYSA-N
Synonym 4-nitrophenyl phenyl sulfide, 4-nitrodiphenyl sulfide, 1-nitro-4-phenylsulfanyl benzene, p-nitrodiphenyl sulfide, p-nitrophenyl phenyl sulfide, 4-nitrodiphenyl thioether, 1-nitro-4-phenylthio benzene, phenyl p-nitrophenylsulfide, benzene, 1-nitro-4-phenylthio, sulfide, p-nitrophenyl phenyl
PubChem CID 13720
IUPAC Name 1-nitro-4-phenylsulfanylbenzene
SMILES [O-][N+](=O)C1=CC=C(SC2=CC=CC=C2)C=C1

Specifications

Melting Point 54°C to 57°C
Quantity 25 g
Formula Weight 231.27
Percent Purity 98%
Chemical Name or Material 4-Nitrophenyl phenyl sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.