missing translation for 'onlineSavingsMsg'
Learn More

(4-Nitrophenyl)phenylamine, TRC

Product Code. 30479617
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479617 500 mg -
30465727 250 mg 250mg
30464952 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479617 Supplier Toronto Research Chemicals Supplier No. TRCN504460500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 836-30-6
Molecular Formula C12 H10 N2 O2
Molecular Weight (g/mol) 214.22
Synonym Benzenamine, 4-nitro-N-phenyl- (9CI, ACI), 4-Nitro-N-phenylbenzenamine (ACI), Diphenylamine, 4-nitro- (8CI), (4-Nitrophenyl)phenylamine, 4-Nitro-N-phenylaniline, 4-Nitrodiphenylamine, N-(4-Nitrophenyl)aniline, N-(4-Nitrophenyl)benzenamine, N-(p-Nitrophenyl)aniline, N-Phenyl-4-nitroaniline, N-Phenyl-4-nitrobenzenamine, N-Phenyl-p-nitroaniline, NSC 33836, p-Nitrodiphenylamine, Phenyl(4-nitrophenyl)amine
IUPAC Name 4-nitro-N-phenylaniline
SMILES [O-][N+](=O)c1ccc(Nc2ccccc2)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H10N2O2/c15-14(16)12-8-6-11(7-9-12)13-10-4-2-1-3-5-10/h1-9,13H
Formula Weight 214.0742
Chemical Name or Material 4-Nitrodiphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.