missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets, Thermo Scientific Chemicals

Product Code. 15450187
Change view
Click to view available options
Quantity:
100 Pc.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450187 100 Pc. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15450187 Supplier Thermo Scientific Alfa Aesar Supplier No. J61401.KN
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Substrate for alkaline and acid phosphatase determinations

4-Nitrophenyl phosphate disodium salt hexahydrate which acts as a chromogenic substrate for the determination of acid and alkaline phosphatases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl phosphate disodium salt hexahydrate which acts as a chromogenic substrate for the determination of acid and alkaline phosphatases.

Solubility
It is soluble in water.

Notes
The tablets should be stored -20°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4264-83-9
Molecular Formula C6H4NNa2O6P
Molecular Weight (g/mol) 263.05
MDL Number MFCD00007319
InChI Key VIYFPAMJCJLZKD-UHFFFAOYSA-L
Synonym pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
PubChem CID 77949
IUPAC Name disodium;(4-nitrophenyl) phosphate
SMILES [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1

Specifications

Color Yellow
Quantity 100 Pc.
Beilstein 5324661
Solubility Information It is soluble in water.
Formula Weight 371.15 (263.05 Anhydrous)
Physical Form Tablets
Chemical Name or Material 4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.