missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%

Product Code. 11985141 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11985141 50 g Glass bottle 50g
10655562 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11985141 Supplier Thermo Scientific Acros Supplier No. 128860500
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, C6H4NNa2O6P.6H2O, CAS Number-333338-18-4 | CAS: 333338-18-4 | C6H4NNa2O6P | 263.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333338-18-4
Molecular Formula C6H4NNa2O6P
Molecular Weight (g/mol) 263.05
MDL Number MFCD00007319
InChI Key VIYFPAMJCJLZKD-UHFFFAOYSA-L
Synonym pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
PubChem CID 77949
IUPAC Name disodium;(4-nitrophenyl) phosphate
SMILES [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1

Specifications

Melting Point >300.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 0.1% max. free 4-nitrophenol (HPLC)
Packaging Glass bottle
Linear Formula O2NC6H4OP(O)(ONa)2·6H2O
Quantity 50 g
Beilstein 06,237
Solubility Information (5% in water) Clear
Formula Weight 371.14
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Nitrophenyl phosphate, disodium salt, hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.