missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl Phosphorodichloridate, TRC

Product Code. 30480324
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480324 10 g -
30471513 5 g 5g
30474221 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30480324 Supplier Toronto Research Chemicals Supplier No. TRCN50453510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 777-52-6
Molecular Formula C6H4Cl2NO4P
Molecular Weight (g/mol) 255.98
Synonym Phosphorodichloridic Acid 4-Nitrophenyl Ester, p-Nitrophenol Phosphorodichloridate, 4-Nitrophenyl Dichlorophosphate, 4-Nitrophenyl Phosphoryl Dichloride, Dichloro(p-nitrophenoxy)phosphine Oxide, O-(4-Nitrophenyl) Phosphorodichloridate, Phosphorodichloridic Acid p-Nitrophenyl Ester, p-Nitrophenyl Dichlorophosphate, p-Nitrophenyl Phosphorodichloridate
IUPAC Name 1-dichlorophosphoryloxy-4-nitrobenzene
SMILES [O-][N+](=O)c1ccc(OP(=O)(Cl)Cl)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H4Cl2NO4P/c7-14(8,12)13-6-3-1-5(2-4-6)9(10)11/h1-4H
Formula Weight 254.9255
Chemical Name or Material 4-Nitrophenyl Phosphorodichloridate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.