missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl sulfate, potassium salt, 99+%

Product Code. 10791915 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10791915 5 g Glass bottle 5g
10542821 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10791915 Supplier Thermo Scientific Acros Supplier No. 227490050
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6217-68-1 | C6H4KNO6S | 257.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6217-68-1
Molecular Formula C6H4KNO6S
Molecular Weight (g/mol) 257.26
MDL Number MFCD00007466
InChI Key BITVAZYUWRLLCN-UHFFFAOYSA-M
Synonym potassium 4-nitrophenyl sulfate, potassium 4-nitrophenyl sulphate, 4-nitrophenyl sulfate potassium salt, potassium p-nitrophenyl sulfate, sulfuric acid, mono 4-nitrophenyl ester, potassium salt, sulfuric acid, mono 4-nitrophenyl ester, potassium salt 1:1, p-nitrophenyl sulfate potassium salt, potassium p-nitrophenyl sulphate, phenol, p-nitro-, hydrogen sulfate ester , potassium salt
PubChem CID 80348
IUPAC Name potassium;(4-nitrophenyl) sulfate
SMILES [K+].[O-][N+](=O)C1=CC=C(OS([O-])(=O)=O)C=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 246.0°C to 250.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 0.05% max. free p-nitrophenol (HPLC)
Packaging Glass bottle
Linear Formula O2NC6H4OSO3K
Quantity 5 g
Beilstein 06, II, 225
Solubility Information Solubility in water: slightly soluble in water.
Formula Weight 257.26
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl sulfate, potassium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.