missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl trifluoromethanesulfonate, 99%

Product Code. 11398446
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398446 1 g 1g
11308456 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398446 Supplier Thermo Scientific Alfa Aesar Supplier No. L17481.03
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 17763-80-3 | C7H4F3NO5S | 271.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17763-80-3
Molecular Formula C7H4F3NO5S
Molecular Weight (g/mol) 271.17
MDL Number MFCD00192354
InChI Key NDTIXHNCNLKURN-UHFFFAOYSA-N
Synonym trifluoromethanesulfonic acid 4-nitrophenyl ester, 4-nitrophenyltriflate, 4-nitrophenyl triflate, acmc-20amc8, 4-nitrophenyl trifluoromethyl sulfonate, 4-nitrophenyl trifluoromethanesulfonate, 1-trifluoromethylsulfonyloxy-4-nitrobenzene, 4-trifluoromethylsulfonyloxy-1-nitrobenzene
PubChem CID 549870
IUPAC Name (4-nitrophenyl) trifluoromethanesulfonate
SMILES [O-][N+](=O)C1=CC=C(OS(=O)(=O)C(F)(F)F)C=C1

Specifications

Melting Point 50°C to 54°C
Quantity 1 g
UN Number UN2923
Beilstein 1998336
Sensitivity Moisture sensitive
Formula Weight 271.17
Percent Purity 99%
Chemical Name or Material 4-Nitrophenyl trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.