Learn More
4-Nitrophthalic anhydride, tech. 90%
Description
4-Nitrophthalic anhydride is used an an intermediate for the synthesis of a benzimidazole PARP inhibitor I (succinate salt) (ABT-472).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Nitrophthalic anhydride is used an an intermediate for the synthesis of a benzimidazole PARP inhibitor I (succinate salt) (ABT-472).
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents. Incompatible with water, bases, strong acids and oxidizing agents.
Specifications
Specifications
| Melting Point | 116°C to 121°C |
| Quantity | 5 g |
| Beilstein | 179682 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Hydrolyzes in water. |
| Formula Weight | 193.11 |
| Percent Purity | 90% |
| Grade | Technical |
| Chemical Name or Material | 4-Nitrophthalic anhydride |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.