missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophthalonitrile, 97%

Product Code. 15496367
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496367 10 g 10g
15486367 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496367 Supplier Thermo Scientific Alfa Aesar Supplier No. H33546.09
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31643-49-9 | C8H3N3O2 | 173.13 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31643-49-9
Molecular Formula C8H3N3O2
Molecular Weight (g/mol) 173.13
MDL Number MFCD00040301
InChI Key NTZMSBAAHBICLE-UHFFFAOYSA-N
Synonym 4-nitrophthalonitrile, 5-nitrobenzene-1,2-dicarbonitrile, 4-nitro-1,2-benzenedicarbonitrile, 1,2-dicyano-4-nitrobenzene, 3,4-dicyanonitrobenzene, phthalonitrile, 4-nitro, 4-nitro-phthalonitrile, 1,2-benzenedicarbonitrile, 4-nitro, 5-nitro-1,2-benzenedicarbonitrile, 4-nitro-o-benzenedinitrile
PubChem CID 97443
IUPAC Name 4-nitrobenzene-1,2-dicarbonitrile
SMILES [O-][N+](=O)C1=CC=C(C#N)C(=C1)C#N

Spécification

Melting Point 136°C to 137°C
Quantity 10 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 173.13
Percent Purity 97%
Chemical Name or Material 4-Nitrophthalonitrile

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.