missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroquinoline-N-oxide, 98%

Product Code. 10460671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10460671 1 g Glass bottle 1g
10002640 250 mg Glass bottle 250mg
10599233 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10460671 Supplier Thermo Scientific Acros Supplier No. 203790010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56-57-5 | C9H6N2O3 | 190.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-57-5
Molecular Formula C9H6N2O3
Molecular Weight (g/mol) 190.16
MDL Number MFCD00006738
InChI Key YHQDZJICGQWFHK-UHFFFAOYSA-N
Synonym 4-nitroquinoline n-oxide, 4-nitroquinoline 1-oxide, 4-nitroquinoline-1-oxide, nitrochin, 4-nitroquinoline-n-oxide, 4-nqo, quinoline, 4-nitro-, 1-oxide, 4-nitroquinoline oxide, 4 nqo, 4-nitroquinolin-1-oxide
PubChem CID 5955
ChEBI CHEBI:16907
IUPAC Name 4-nitro-1-oxidoquinolin-1-ium
SMILES [O-][N+](=O)C1=C2C=CC=CC2=[N+]([O-])C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 154°C to 156°C
Color Brown to Ochre-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in acetone,dmso,dmf
Formula Weight 190.16
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4-Nitroquinoline-N-oxide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Nitroquinoline-N-oxide
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.