missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrothiobenzamide, 97%

Product Code. 15498657
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498657 5 g 5g
15488657 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498657 Supplier Thermo Scientific Alfa Aesar Supplier No. H52140.06
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26060-30-0 | C7H6N2O2S | 182.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26060-30-0
Molecular Formula C7H6N2O2S
Molecular Weight (g/mol) 182.197
MDL Number MFCD06150000
InChI Key MDBQPBMIZPCKAJ-UHFFFAOYSA-N
Synonym 4-nitrobenzothioamide, 4-nitro-thiobenzamide, 4-nitrobenzene-1-carbothioamide, 4-nitrothiobenzamide, benzenecarbothioamide,4-nitro, benzenecarbothioamide, 4-nitro, p-nitrothiobenzamide, 4-nitro-benzenecarbothioamide, 4-nitrobenzene-carbothioamide, amino 4-nitrophenyl methane-1-thione
PubChem CID 3000564
IUPAC Name 4-nitrobenzenecarbothioamide
SMILES C1=CC(=CC=C1C(=S)N)[N+](=O)[O-]

Specifications

Quantity 5 g
Formula Weight 182.2
Percent Purity 97%
Chemical Name or Material 4-Nitrothiobenzamide

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.