missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrothiophenol (Contains Dimer), TRC

Product Code. 30483680
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483680 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483680 Supplier Toronto Research Chemicals Supplier No. TRCN5655205G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Nitrothiophenol
CAS 1849-36-1
Molecular Formula C6H5NO2S
Quantity 5 g
Synonym 4-Nitrobenzenethiol, 4-Nitrophenyl Mercaptan, NSC 128182, p-Nitrobenzenethiol, p-Nitrophenyl Mercaptan, p-Nitrothiophenol
InChI Formula InChI=1S/C6H5NO2S/c8-7(9)5-1-3-6(10)4-2-5/h1-4,10H
SMILES C1=C(C=CC(=C1)S)[N+](=O)[O-]
IUPAC Name 4-nitrobenzenethiol
Molecular Weight (g/mol) 155.17
Formula Weight 155.0041
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.