missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrotoluene-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 4-Nitrotoluene ring-d4 |
| CAS | 1276197-16-0 |
| Molecular Formula | C7 2H4 H3 N O2 |
| Quantity | 0.25 g |
| Synonym | Benzene-1,2,4,5-d4, 3-methyl-6-nitro-, 4-Nitrotoluene-2,3,5,6-d4, 4-Nitrotoluene (ring-D4), 1-Methyl-4-nitro-2,3,5,6-tetradeuteriobenzene |
| InChI Formula | InChI=1 S/C7H7NO2/c1-6-2-4-7(5-3-6)8(9)10/h2-5 H,1H3/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1C)[N+](=O)[O-] |
| IUPAC Name | 1,2,4,5-tetradeuterio-3-methyl-6-nitrobenzene |
| Molecular Weight (g/mol) | 141.16 |
| Formula Weight | 141.0728 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction