missing translation for 'onlineSavingsMsg'
Learn More

4-Nonyl Phenol-13C6 Monoethoxylate, TRC

Product Code. 30132449
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132449 5 mg 5mg
30011205 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132449 Supplier Toronto Research Chemicals Supplier No. TRCN6504525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1346600-52-9
Molecular Formula C1113C6H28O2
Molecular Weight (g/mol) 270.36
Synonym 2-(4-Nonylphenoxy)ethanol-13C6, 2-(p-Nonylphenoxy)ethanol-13C6, 2-(4-Nonylphenoxy)ethyl Alcohol-13C6, 2-(p-Nonylphenoxy)ethanol-13C6, 4-Nonylphenol Monoethoxylate-13C6, Ethylene Glycol Mono-p-nonylphenyl Ether-13C6, Ethylene Glycol p-Nonylphenyl Ether-13C6;
SMILES OCCO[13C]1=[13CH][13CH]=[13C](CCCCCCCCC)[13CH]=[13CH]1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C17H28O2/c1-2-3-4-5-6-7-8-9-16-10-12-17(13-11-16)19-15-14-18/h10-13,18H,2-9,14-15H2,1H3/i10+1,11+1,12+1,13+1,16+1,17+1
Formula Weight 270.2291
Chemical Name or Material 4-Nonyl Phenol-13C6 Monoethoxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.