missing translation for 'onlineSavingsMsg'
Learn More

4-Nonyl Phenol Diethoxylate, TRC

Product Code. 30481458
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481458 50 mg 50mg
30477772 5 mg -
30464758 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481458 Supplier Toronto Research Chemicals Supplier No. TRCN65044050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20427-84-3
Molecular Formula C19 H32 O3
Molecular Weight (g/mol) 308.46
Synonym 2-[2-(4-Nonylphenoxy)ethoxy]ethanol, 2-[2-(p-Nonylphenoxy)ethoxy]ethanol, 4-Nonylphenol diethoxylate, 4-n-Nonylphenol diethoxylate, Diethylene glycol mono-p-nonylphenyl ether, Diethylene glycol p-nonylphenyl ether, EON 2
IUPAC Name 2-[2-(4-nonylphenoxy)ethoxy]ethanol
SMILES CCCCCCCCCc1ccc(OCCOCCO)cc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H32O3/c1-2-3-4-5-6-7-8-9-18-10-12-19(13-11-18)22-17-16-21-15-14-20/h10-13,20H,2-9,14-17H2,1H3
Formula Weight 308.2351
Percent Purity >95%
Chemical Name or Material 4-n-Nonylphenol-diethoxylate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.