missing translation for 'onlineSavingsMsg'
Learn More

4-O-Beta-Galactopyranosyl-D-mannopyranoside, TRC

Product Code. 30010621
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010621 500 mg 500mg
30009588 10 mg 10mg
30009134 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010621 Supplier Toronto Research Chemicals Supplier No. TRCG154850500MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20869-27-6
Molecular Formula C12 H22 O11
Molecular Weight (g/mol) 342.3
Synonym D-Mannopyranose, 4-O-β-D-galactopyranosyl-, Mannopyranose, 4-O-β-D-galactopyranosyl-, D- (8CI), Epilactose
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3-,4-,5+,6+,7-,8+,9-,10-,11?,12+/m1/s1
Formula Weight 342.1162
Percent Purity >95%
Chemical Name or Material 4-O-beta-Galactopyranosyl-D-mannopyranoside
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.