missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Octylbenzoic Acid, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 4-Octylbenzoic Acid |
| CAS | 3575-31-3 |
| Molecular Formula | C15H22O2 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C15H22O2/c1-2-3-4-5-6-7-8-13-9-11-14(12-10-13)15(16)17/h9-12H,2-8H2,1H3,(H,16,17) |
| SMILES | CCCCCCCCc1ccc(cc1)C(=O)O |
| IUPAC Name | 4-octylbenzoic acid |
| Molecular Weight (g/mol) | 234.34 |
| Formula Weight | 234.162 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction