missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-4-(4-toluidino)but-2-enoic acid, 97%, Thermo Scientific™

Product Code. 10009281
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10009281 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10009281 Supplier Thermo Scientific Maybridge Supplier No. S01331EA
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 37904-03-3
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H11NO3
Linear Formula CH3C6H4NHC(O)CH=CHCO2H
MDL Number MFCD00020480
Quantity 10 g
Synonym 2z-3-4-methylphenyl carbamoyl prop-2-enoic acid, 4-oxo-4-4-toluidino but-2-enoic acid, 4-4-methylphenyl amino-4-oxobut-2-enoic acid, n-p-tolyl-maleamic acid, n-p-tolylmaleamidic acid, maleanilic acid, p-methyl, n-4-methylphenyl maleamic, z-4-oxo-4-p-tolylamino but-2-enoic acid, z-4-4-methylanilino-4-oxobut-2-enoic acid
InChI Key VLWFSWMZGGZHGD-SREVYHEPSA-N
SMILES CC1=CC=C(C=C1)NC(=O)C=CC(=O)O
IUPAC Name (Z)-4-(4-methylanilino)-4-oxobut-2-enoic acid
Molecular Weight (g/mol) 205.213
PubChem CID 5356452
Formula Weight 205.21
Percent Purity 97%
Chemical Name or Material 4-Oxo-4-(4-toluidino)but-2-enoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Oxo-4-(4-toluidino)but-2-enoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.