missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo Cyclophosphamide, TRC

Product Code. 30007889
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007889 1 mg 1mg
30004520 5 mg 5mg
30007102 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007889 Supplier Toronto Research Chemicals Supplier No. TRCO8499001MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27046-19-1
Molecular Formula C7 H12 Cl2 N O4 P
Molecular Weight (g/mol) 276.05
IUPAC Name 2-[bis(2-chloroethyl)amino]-2-oxo-1,3,2λ^{5}-dioxaphosphinan-5-one
SMILES ClCCN(CCCl)P1(=O)OCC(=O)CO1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C7H12Cl2NO4P/c8-1-3-10(4-2-9)15(12)13-5-7(11)6-14-15/h1-6H2
Formula Weight 274.9881
Percent Purity >95%
Chemical Name or Material 4-Oxo Cyclophosphamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.