missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-L-proline hydrobromide, 90+%

Product Code. 15474128
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15474128 1 g 1g
15484128 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474128 Supplier Thermo Scientific Alfa Aesar Supplier No. H57691.03
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75776-67-9 | C5H8BrNO3 | 210.027 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75776-67-9
Molecular Formula C5H8BrNO3
Molecular Weight (g/mol) 210.027
MDL Number MFCD12024572
InChI Key SFSROLZHCPCODJ-WCCKRBBISA-N
Synonym 4-keto-l-proline hydrobromide, s-4-oxopyrrolidine-2-carboxylic acid hydrobromide, 4-keto-l-prolinehydrobromide, 4-oxo-l-proline hydrobromide, 4-oxoproline hydrobromide, l-proline, 4-oxo-, hydrobromide, s-4-keto-proline hydrobromide, pubchem14289, 4-keto-l-proline, hydrobromide
PubChem CID 12773661
IUPAC Name (2S)-4-oxopyrrolidine-2-carboxylic acid;hydrobromide
SMILES C1C(NCC1=O)C(=O)O.Br

Specifications

Melting Point 134°C to 140°C
Quantity 1 g
Formula Weight 210.03
Optical Rotation −32° (c=0.5 in Methanol)
Sensitivity Moisture sensitive
Percent Purity ≥90%
Chemical Name or Material 4-Oxo-L-proline hydrobromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.