missing translation for 'onlineSavingsMsg'
Learn More

4-Pentylbenzonitrile, 97%, Thermo Scientific™

Product Code. 10324752
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10324752 10 g Amber Glass Bottle 10g
10191242 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10324752 Supplier Thermo Scientific Maybridge Supplier No. BTB12834EA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10270-29-8
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.26
MDL Number MFCD00218832
InChI Key LGPQFJXPPKNHJU-UHFFFAOYSA-N
Synonym 4-n-pentylbenzonitrile, 4-pentylbenzenecarbonitrile, p-amylbenzonitrile, p-n-pentylbenzonitrile, benzonitrile, 4-pentyl
PubChem CID 264951
IUPAC Name 4-pentylbenzonitrile
SMILES CCCCCC1=CC=C(C=C1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3(CH2)4C6H4CN
Quantity 10 g
Formula Weight 173.26
Percent Purity 97%
Chemical Name or Material 4-Pentylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.