missing translation for 'onlineSavingsMsg'
Learn More

4-Pentyn-1-ol, 95%

Product Code. 10721181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10721181 25 g Glass bottle 25g
10608231 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10721181 Supplier Thermo Scientific Acros Supplier No. 330430250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5390-04-5 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5390-04-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00002974
InChI Key CRWVOXFUXPYTRK-UHFFFAOYSA-N
Synonym 6-bromoveratraldehyde, 6-bromo-3,4-dimethoxybenzaldehyde, benzaldehyde, 2-bromo-4,5-dimethoxy, 6-bromo veratraldehyde, 2-bromo-4,5-dimethoxybenzalehyde, 4,5-dimethoxy-2-bromobenzaldehyde, 6-bromoveratral, pubchem8209, veratraldehyde, 6-bromo, acmc-2097hh
PubChem CID 79351
SMILES OCCCC#C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 0.9100g/mL
Boiling Point 154°C to 155°C
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula HC∽C(CH2)3OH
Refractive Index 1.4430 to 1.447
Quantity 25 g
Specific Gravity 0.91
IUPAC Name pent-4-yn-1-ol
Formula Weight 84.12
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Pentyn-1-ol, 95%
Show More Show Less

RUO – Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.