missing translation for 'onlineSavingsMsg'
Learn More

4-Pentyn-2-ol, 97+%

Product Code. 10493385 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10493385 5 g Glass bottle 5g
10320494 1 g Glass bottle 1g
10501544 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10493385 Supplier Thermo Scientific Supplier No. 392670050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2117-11-5 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2117-11-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00004555
InChI Key JTHLRRZARWSHBE-UHFFFAOYNA-N
Synonym 4-pentyn-2-ol, 4-pentyne-2-ol, +--4-pentyn-2-ol, r,s-4-pentyn-2-ol, 4-hydroxypentyne, pentyne-4-ol, 4-pentin-2-ol, 1-methyl-3-butyn-1-ol, +/--4-pentyn-2-ol
PubChem CID 92915
IUPAC Name pent-4-yn-2-ol
SMILES CC(CC#C)O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8960g/mL
Boiling Point 126°C to 127°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula CHCCH2CH(OH)CH3
Refractive Index 1.4360 to 1.438
Quantity 5 g
Specific Gravity 0.896
Formula Weight 84.12
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material 4-Pentyn-2-ol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Pentyn-2-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.