missing translation for 'onlineSavingsMsg'
Learn More

4-Pentynoic acid, 95%

Product Code. 10533191 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10533191 5 g Glass bottle 5g
10226700 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10533191 Supplier Thermo Scientific Acros Supplier No. 213730050
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6089-09-4 | C5H6O2 | 98.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6089-09-4
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.1
MDL Number MFCD00004407
InChI Key MLBYLEUJXUBIJJ-UHFFFAOYSA-M
Synonym 4-pentynoic acid, propargylacetic acid, pent-4-yn-1-oic acid, 4-pentynoicacid, pent4-ynoic acid, zlchem 796, pubchem3491, pent-4-yn-oic acid, acmc-1az7d
PubChem CID 22464
IUPAC Name pent-4-ynoic acid
SMILES C#CCCC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 54.0°C to 58.0°C
Color White to Beige
Boiling Point 110.0°C (30.0 mmHg)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CCH2CH2CO2H
Quantity 5 g
Beilstein 02, 481
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone and,methyl-t.-buthylether
Formula Weight 98.1
Percent Purity 95%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 4-Pentynoic acid, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Pentynoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.