missing translation for 'onlineSavingsMsg'
Learn More

4-Pentynoic Acid, TRC

Product Code. 30423841
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423841 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423841 Supplier Toronto Research Chemicals Supplier No. TRCP2849001G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 4-Pentynoic Acid
CAS 6089-09-4
Molecular Formula C5H6O2
Quantity 1 g
Synonym Propargylacetic Acid
InChI Formula InChI=1S/C5H6O2/c1-2-3-4-5(6)7/h1H,3-4H2,(H,6,7)
SMILES OC(=O)CCC#C
IUPAC Name pent-4-ynoic acid
Molecular Weight (g/mol) 98.1
Formula Weight 98.0368
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.