missing translation for 'onlineSavingsMsg'
Learn More

4-Phenanthrol-d9, TRC

Product Code. 30424974
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2.5 mg
Unit Size:
0.5mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424974 0.5 mg 0.5mg
30422991 1 mg 1mg
30422872 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30424974 Supplier Toronto Research Chemicals Supplier No. TRCP2950170.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 922510-21-2
Molecular Formula C14HD9O
Molecular Weight (g/mol) 203.28
Synonym Phenanthren-1,2,3,5,6,7,8,9,10-d9-4-ol, 4-Phenanthrenol-d9, 4-Phenanthrol-d9, 4-Hydroxyphenanthrene-d9, NSC 171284-d9
IUPAC Name phenanthren-d9-4-ol
SMILES [2H]C1=C([2H])C([2H])=C([2H])C2=C1C([2H])=C([2H])C3=C([2H])C([2H])=C([2H])C(O)=C32

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C14H10O/c15-13-7-3-5-11-9-8-10-4-1-2-6-12(10)14(11)13/h1-9,15H/i1D,2D,3D,4D,5D,6D,7D,8D,9D
Formula Weight 203.13
Percent Purity >95%
Chemical Name or Material 4-Phenanthrol-d9
Grade HPLC
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.