missing translation for 'onlineSavingsMsg'
Learn More

4-(Phenoxymethyl)phenylboronic acid, 97%, Thermo Scientific™

Product Code. 10557294
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10557294 1 g Amber Glass Bottle 1g
10183904 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10557294 Supplier Thermo Scientific Maybridge Supplier No. CC63712DA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 397843-61-7
Molecular Formula C13H13BO3
Molecular Weight (g/mol) 228.05
MDL Number MFCD09966155
InChI Key IXDHTLPOOZIZJX-UHFFFAOYSA-N
Synonym 4-phenoxymethyl phenylboronic acid, 4-phenoxymethyl phenyl boronic acid, acmc-1aegd, 4-phenoxymethyl phenyl boronicacid
PubChem CID 10955308
IUPAC Name [4-(phenoxymethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=C(COC2=CC=CC=C2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 228.05
Percent Purity 97%
Chemical Name or Material 4-(Phenoxymethyl)phenylboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.