missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1,2,3,4-tetrahydroisoquinoline Hydrochloride, TRC

Product Code. 30409309
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409309 25 mg 25mg
30415890 10 mg -
30427128 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30409309 Supplier Toronto Research Chemicals Supplier No. TRCP33693525MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6109-35-9
Molecular Formula C15 H15 N . Cl H
Molecular Weight (g/mol) 245.75
IUPAC Name 4-phenyl-1,2,3,4-tetrahydroisoquinoline;hydrochloride
SMILES Cl.C1NCc2ccccc2C1c3ccccc3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H15N.ClH/c1-2-6-12(7-3-1)15-11-16-10-13-8-4-5-9-14(13)15;/h1-9,15-16H,10-11H2;1H
Formula Weight 245.0971
Chemical Name or Material 4-Phenyl-1,2,3,4-tetrahydroisoquinoline Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.