missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1-piperidin-4-yl-1,3-dihydro-2H-imidazol-2-one, Thermo Scientific™

Product Code. 10381650
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10381650 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10381650 Supplier Thermo Scientific Maybridge Supplier No. MAY00232DA
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 205058-28-2
Packaging Amber Glass Bottle
Molecular Formula C14H17N3O
Quantity 1 g
Synonym 4-phenyl-1-4-piperidinyl-4-imidazolin-2-one, 4-phenyl-1-piperidin-4-yl-1,3-dihydro-2h-imidazol-2-one, 4-phenyl-1-piperidin-4-yl-1h-imidazol-2 3h-one, 4-phenyl-1-piperidin-4-yl-3h-imidazol-2-one, 4-phenyl-1-piperidin-4-yl-2,3-dihydro-1h-imidazol-2-one, 4-phenyl-1-4-piperidyl-4-imidazolin-2-one, acmc-1chii, 4-phenyl-piperidin-4-yl-1h-imidazol-2 3h-one, 1,3-dihydro-1-4-piperidinyl-4-phenyl-2 2h-imidazolone, 2h-imidazol-2-one, 1,3-dihydro-4-phenyl-1-4-piperidinyl
InChI Key OPCVKWIYGHICKG-UHFFFAOYSA-N
SMILES C1CNCCC1N2C=C(NC2=O)C3=CC=CC=C3
IUPAC Name 5-phenyl-3-piperidin-4-yl-1H-imidazol-2-one
Molecular Weight (g/mol) 243.31
PubChem CID 11345546
Formula Weight 243.31
Percent Purity 97%
Chemical Name or Material 4-Phenyl-1-piperidin-4-yl-1, 3-dihydro-2H-imidazol-2-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.